Novus Biologicals
Manufacturer Code:NBP272930AF647
Catalog # N272930A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E10 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-274 of human NEDD8 Activating Enzyme (APPBP1/UBA3) (NP_001018170) produced in E.coli. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: A-116A10.1 amyloid beta precursor protein binding protein 1 amyloid beta precursor protein binding protein 1 59kDa Amyloid beta precursor protein-binding protein 1 59 kDa amyloid beta precursor protein-binding protein 1 59kD Amyloid protein-binding protein 1 APPBP1APP-BP1 NEDD8 activating enzyme E1 subunit 1 NEDD8-activating enzyme E1 regulatory subunit NEDD8-activating enzyme E1 subunit protooncogene protein 1 Proto-oncogene protein 1 ula-1; amyloid beta precursor protein binding protein 1, 59kDa; Amyloid beta precursor protein-binding protein 1, 59 kDa; amyloid beta precursor protein-binding protein 1, 59kD; Amyloid protein-binding protein 1; APP-BP1; NEDD8-activating enzyme E1 regulatory subunit; NEDD8-activating enzyme E1 subunit; Proto-oncogene protein 1; protooncogene protein 1
Gene Aliases: A-116A10.1; APPBP1; HPP1; NAE1; ula-1
UniProt ID: (Human) Q13564
Entrez Gene ID: (Human) 8883
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.