Invitrogen
Manufacturer Code:PA579726
Catalog # PIPA579726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human NEDD4-2 recombit protein (Position: N703-D975). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase NEDD4-like; E3 ubiquitin-protein ligase NEDD4-like HECT-type E3 ubiquitin transferase NED4L NEDD4.2 Nedd4-2 NEDD4L KIAA0439 NEDL3; HECT-type E3 ubiquitin transferase NED4L; Nedd4-2; NEDD4.2; neural precursor cell expressed, developmentally down-regulated gene 4b; ubiquitin-protein ligase Rsp5
Gene Aliases: 1300012C07Rik; hNEDD4-2; KIAA0439; NEDD4-2; NEDD4.2; Nedd4b; NEDD4L; NEDL3; RSP5
UniProt ID: (Human) Q96PU5
Entrez Gene ID: (Human) 23327, (Rat) 291553, (Mouse) 83814
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.