Invitrogen
Manufacturer Code:PA542310
Catalog # PIPA542310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NEDD4L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase NEDD4-like; HECT-type E3 ubiquitin transferase NED4L; Nedd4-2; NEDD4.2; nedd4b; Neural precursor cell expressed developmentally down-regulated 4-like E3 ubiquitin protein ligase FLJ33870 KIAA0439 RSP5 hNedd4-2 NEDD4-2 NEDD4.2 hNEDD4-2; neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4-like; neural precursor cell expressed, developmentally down-regulated gene 4b; ubiquitin-protein ligase Nedd4-2; ubiquitin-protein ligase Rsp5
Gene Aliases: 1300012C07Rik; hNEDD4-2; KIAA0439; NEDD4; NEDD4-2; NEDD4.2; Nedd4b; NEDD4L; NEDL3; RSP5
UniProt ID: (Human) Q96PU5
Entrez Gene ID: (Human) 23327, (Dog) 476189, (Rabbit) 100353099, (Bovine) 510003, (Horse) 100054395, (Rat) 291553, (Guinea pig) 100301562, (Mouse) 83814, (Zebrafish) 559634
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.