Invitrogen
Manufacturer Code:PA517463
Catalog # PIPA517463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human NEDD4 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 53 protein; E3 ubiquitin-protein ligase NEDD4; E3 ubiquitin-protein ligase NEDD4 NEDD-4 OTTHUMP00000217840 OTTHUMP00000217841 OTTHUMP00000217842 cell proliferation-inducing gene 53 protein neural precursor cell expressed developmentally down-regulated protein 4 receptor-potentiating factor 1; HECT-type E3 ubiquitin transferase NEDD4; NEDD-4; Neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4A; neural precursor cell expressed, developmentally down-regulted gene 4; receptor-potentiating factor 1
Gene Aliases: AA959633; AL023035; AU019897; E430025J12Rik; KIAA0093; mKIAA0093; Nedd-4; NEDD4; NEDD4-1; Nedd4a; PIG53; RPF1
UniProt ID: (Human) B4E2R7, (Rat) Q62940, (Mouse) P46935
Entrez Gene ID: (Human) 4734, (Rat) 25489, (Mouse) 17999
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.