Invitrogen
Manufacturer Code:PA570092
Catalog # PIPA570092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse NECAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adaptin ear-binding clathrin-associated protein; Adaptin ear-binding coat-associated protein 1; adaptin-ear-binding coat-associated protein 1; brain enriched NECAP2 similar protein; clathrin-coated vesicle coat protein; EIEE21 adaptin ear-binding coat-associated protein 1; NECAP endocytosis-associated protein 1; NECAP-1; snoRNA MBI-129
Gene Aliases: 1200016B17Rik; AI747569; EIEE21; mFLJ00061; NECAP1; RGD1306053; zgc:73124
UniProt ID: (Bovine) Q3T093, (Human) Q8NC96, (Rat) P69682, (Mouse) Q9CR95
Entrez Gene ID: (Dog) 486705, (Bovine) 504449, (Human) 25977, (Rabbit) 100344988, (Horse) 100060780, (Guinea pig) 100725574, (Rat) 312694, (Mouse) 67602, (Zebrafish) 393695
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.