Proteintech
Manufacturer Code:179061AP150UL
Catalog # 50-173-1866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NECAP1 Fusion Protein Ag12283 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adaptin ear-binding clathrin-associated protein; Adaptin ear-binding coat-associated protein 1; adaptin-ear-binding coat-associated protein 1; brain enriched NECAP2 similar protein; clathrin-coated vesicle coat protein; NECAP 1 NECAP endocytosis associated 1 NECAP1; NECAP endocytosis-associated protein 1; NECAP-1; snoRNA MBI-129
Gene Aliases: 1200016B17Rik; AI747569; EIEE21; mFLJ00061; NECAP1; RGD1306053
UniProt ID: (Human) Q8NC96, (Rat) P69682, (Mouse) Q9CR95
Entrez Gene ID: (Human) 25977, (Rat) 312694, (Mouse) 67602
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.