Invitrogen
Manufacturer Code:PA539020
Catalog # PIPA539020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human NECAB3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amyloid beta (A4) precursor protein-binding family A member 1 binding protein; amyloid beta (A4) precursor protein-binding, family A, member 1 binding protein; amyloid beta (A4) precursor protein-binding, family A, member 2 binding protein; amyloid beta A4 protein-binding family A member 2-binding protein; Amyloid-beta A4 protein-binding family A member 2-binding protein; APBA2BP EFCBP3 NIP1 STIP3 SYTIP2 XB51 dJ63M2.4 dJ63M2.5 EF-hand calcium binding protein 3 N-terminal EF-hand calcium-binding protein 3 Nek2-interacting protein 1 X11L-binding protein 51 amyloid beta (A4) precursor protein-binding family A member 2 binding protein amyloid beta A4 protein-binding family A memb; EF-hand calcium binding protein 3; mXB51; N-terminal EF-hand calcium-binding protein 3; Nek2-interacting protein 1; Neuronal calcium-binding protein 3; neuronal calcium-binding protein NECAB3; synaptotagmin interacting protein 2; synaptotagmin interacting protein STIP3; X11L-binding protein 51
Gene Aliases: 2900010M17Rik; AI853434; APBA2BP; dJ63M2.4; dJ63M2.5; EFCBP3; NECAB3; NIP1; STIP3; SYTIP2; XB51
UniProt ID: (Human) Q96P71, (Mouse) Q9D6J4
Entrez Gene ID: (Human) 63941, (Mouse) 56846
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.