Invitrogen
Manufacturer Code:MA524617
Catalog # PIMA524617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL0524 |
Immunogen | Recombit protein corresponding to Human NECAB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EF hand calcium binding protein 2; EF-hand calcium binding protein 2; EF-hand calcium-binding protein 2; N-terminal EF-hand calcium-binding protein 2; neuronal calcium binding 2; Neuronal calcium-binding protein 2; Stip-2; Synaptotagmin-interacting protein 2
Gene Aliases: EFCBP2; NECAB2
UniProt ID: (Human) Q7Z6G3
Entrez Gene ID: (Human) 54550
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.