Novus Biologicals
Manufacturer Code:NBP272923AF532
Catalog # NP272923A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G8 |
Immunogen | Full length recombinant protein of human NECAB1 (NP_071746) produced in HEK293T cells. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EF hand calcium binding protein 1; EF hand calcium binding protein 1 EFCBP1 EF-hand calcium binding protein 1 EF-hand calcium-binding protein 1 neuronal calcium binding protein Neuronal calcium-binding protein 1 N-terminal EF-hand calcium binding protein 1 N-terminal EF-hand calcium-binding protein 1 STIP-1 synaptotagmin interacting protein 1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; N-terminal EF-hand calcium-binding protein 1; neuronal calcium binding protein; Neuronal calcium-binding protein 1; synaptotagmin interacting protein 1
Gene Aliases: EFCBP1; NECAB1; STIP-1
UniProt ID: (Human) Q8N987
Entrez Gene ID: (Human) 64168
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.