Invitrogen
Manufacturer Code:PA554849
Catalog # PIPA554849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NECAB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EF hand calcium binding protein 1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; N-terminal EF-hand calcium-binding protein 1; N-terminal EF-hand calcium-binding protein 1 EF-hand calcium-binding protein 1 Neuronal calcium-binding protein 1 NECAB1 EFCBP1; neuronal calcium binding protein; Neuronal calcium-binding protein 1; synaptotagmin interacting protein 1
Gene Aliases: 1700003H21Rik; EFCBP1; NECAB1; STIP-1
UniProt ID: (Human) Q8N987, (Mouse) Q8BG18
Entrez Gene ID: (Human) 64168, (Mouse) 69352
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.