Invitrogen
Manufacturer Code:PA519343
Catalog # PIPA519343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SRGEYVVAKLD) corresponding to the internal amino acids of NDUFS7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CI-20kD; complex I 20kDa subunit; complex I, mitochondrial respiratory chain, 20-KD subunit; Complex I-20kD; NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 20 kDa subunit; NADH:ubiquinone oxidoreductase PSST subunit; NDUFS7 NADH dehydrogenase (ubiquinone) Fe-S protein 7 20kDa (NADH-coenzyme Q reductase) CI-20KD FLJ45860 FLJ46880 MGC120002 MY017 PSST NADH dehydrogenase &lsqbubiquinone] iron-sulfur protein 7 mitochondrial NADH-coenzyme Q reductase NADH-ubiquinone oxidoreductase Fe-S protein 7 NADH:ubiquinone oxidoreductase PSST subunit complex I mitochondrial respiratory chain 20-KD subunit complex I-20kD; PSST subunit
Gene Aliases: CI-20; CI-20KD; MY017; NDUFS7; PSST
UniProt ID: (Human) O75251
Entrez Gene ID: (Human) 374291
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.