Invitrogen
Manufacturer Code:PA572499
Catalog # PIPA572499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 131-160 amino acids from the C-terminal region of human NDUFS4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AQDQ CI-18 CI-18 kDa CI-AQDQ NADH dehydrogenase (ubiquinone) iron-sulfur protein 4 NADH dehydrogenase &lsqbubiquinone] iron-sulfur protein 4 mitochondrial NADH-coenzyme Q reductase 18-KD NADH-ubiquinone oxidoreductase 18 kDa subunit complex I 18kDa subunit complex I-18 kDa complex I-AQDQ mitochondrial respiratory chain complex I (18-KD subunit); CI-18 kDa; CI-AQDQ; complex I 18kDa subunit; Complex I-18 kDa; Complex I-AQDQ; mitochondrial respiratory chain complex I (18-KD subunit); NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial; NADH-ubiquinone oxidoreductase 18 kDa subunit
Gene Aliases: AQDQ; CI-18; CI-18 kDa; CI-AQDQ; NDUFS4
UniProt ID: (Human) O43181
Entrez Gene ID: (Human) 4724
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.