Invitrogen
Manufacturer Code:PA5103951
Catalog # PIPA5103951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NDUFS4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6720411N02Rik Aqdq C1-18k CI-18 CI-18 kDa CI-AQDQ complex I 18kDa subunit complex I-18 kDa Complex I-AQDQ mitochondrial respiratory chain complex I (18-KD subunit) NADH dehydrogenase (ubiquinone) Fe-S protein 4 NADH dehydrogenase (ubiquinone) Fe-S protein 4 18kDa (NADH-coenzyme Q reductase) NADH dehydrogenase [ubiquinone] iron-sulfur protein 4 mitochondrial NADH:ubiquinone oxidoreductase subunit S4 NADH-ubiquinone oxidoreductase 18 kDa subunit Ndufs4; CI-18 kDa; CI-AQDQ; complex I 18kDa subunit; Complex I-18 kDa; Complex I-AQDQ; mitochondrial respiratory chain complex I (18-KD subunit); NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial; NADH-ubiquinone oxidoreductase 18 kDa subunit
Gene Aliases: 6720411N02Rik; AQDQ; C1-18k; CI-18; CI-18 kDa; CI-AQDQ; NDUFS4
UniProt ID: (Human) O43181, (Mouse) Q9CXZ1, (Rat) Q5XIF3
Entrez Gene ID: (Human) 4724, (Mouse) 17993, (Rat) 499529
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.