Abnova
Manufacturer Code:H00004722D01
Catalog # 89-002-904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NDUFS3 (NP_004542.1 1 a.a. ~ 264 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human NDUFS3 protein.
For Research Use Only
Protein Aliases: CI-30kD; complex I 30kDa subunit; Complex I-30kD; NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH-ubiquinone oxidoreductase 30 kDa subunit
Gene Aliases: CI-30; NDUFS3
UniProt ID: (Human) O75489
Entrez Gene ID: (Human) 4722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.