Invitrogen
Manufacturer Code:PA522364
Catalog # PIPA522364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 200 and 430 of NDUFS2 (Uniprot IDNo.O75306) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CI-49 CI-49kD NADH dehydrogenase &lsqbubiquinone] iron-sulfur protein 2 mitochondrial NADH-ubiquinone oxidoreductase 49 kDa subunit NADH-ubiquinone oxidoreductase NDUFS2 subunit complex 1 mitochondrial respiratory chain 49-KD subunit complex I 49kDa subunit complex I-49kD; CI-49kD; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; Complex I-49kD; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit
Gene Aliases: AL033311; CI-49; NDUFS2
UniProt ID: (Human) O75306, (Mouse) Q91WD5
Entrez Gene ID: (Human) 4720, (Mouse) 226646
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.