Invitrogen
Manufacturer Code:PA519342
Catalog # PIPA519342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SRGVRQWQPDVE) corresponding to the internal amino acids of NDUFS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CI-49kD; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; Complex I-49kD; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; NDUFS2 NADH dehydrogenase (ubiquinone) Fe-S protein 2 49kDa (NADH-coenzyme Q reductase) NADH dehydrogenase (ubiquinone) Fe-S protein 2 NADH-ubiquinone oxidoreductase NDUFS2 subunit complex 1 mitochondrial respiratory chain 49-KD subunit; ndufs2 {ECO:0000312|EMBL:AAH82067.1}
Gene Aliases: AL033311; CI-49; NDUFS2
UniProt ID: (Human) O75306, (Mouse) Q91WD5, (Rat) Q641Y2
Entrez Gene ID: (Human) 4720, (Mouse) 226646, (Rat) 289218
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.