Invitrogen
Manufacturer Code:PA5112278
Catalog # PIPA5112278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human NDUFS2 fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AL033311 CI-49 CI-49kD comp complex 1 mitochondrial respiratory chain 49-KD subunit complex I 49kDa subunit Complex I-49kD EMBL:AAH82067.1} NADH dehydrogenase (ubiquinone) Fe-S protein 2 NADH dehydrogenase (ubiquinone) Fe-S protein 2 49kDa (NADH-coenzyme Q reductase) NADH dehydrogenase [ubiquinone] iron-sulfur protein 2 mitochondrial NADH:ubiquinone oxidoreductase core subunit S2 NADH-coenzyme Q reductase NADH-ubiquinone oxidoreductase 49 kDa subunit NADH-ubiquinone oxidoreductase NDUFS2 subunit Ndufs2 ndufs2 {ECO:0000312; CI-49kD; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; Complex I-49kD; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; unnamed protein product
Gene Aliases: CI-49; NDUFS2
UniProt ID: (Human) O75306
Entrez Gene ID: (Cynomolgus monkey) 101925833, (Human) 4720
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.