Novus Biologicals
Manufacturer Code:NBP152061
Catalog # NBP152061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SRGVRQWQPDVE corresponding to internal region according to NP_001159631.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CI-49 CI-49kD complex 1 mitochondrial respiratory chain 49-KD subunit complex I 49kDa subunit Complex I-49kD EC 1.6.5.3 EC 1.6.99.3 EC 1.6.99.5 NADH dehydrogenase (ubiquinone) Fe-S protein 2 (49kD) (NADH-coenzyme Qreductase) NADH dehydrogenase (ubiquinone) Fe-S protein 2 49kDa (NADH-coenzyme Qreductase) NADH dehydrogenase [ubiquinone] iron-sulfur protein 2 mitochondrial NADH-ubiquinone oxidoreductase 49 kDa subunit NADH-ubiquinone oxidoreductase NDUFS2 subunit; CI-49kD; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; Complex I-49kD; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit
Gene Aliases: CI-49; NDUFS2
UniProt ID: (Human) O75306
Entrez Gene ID: (Human) 4720
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.