Invitrogen
Manufacturer Code:PA528220
Catalog # PIPA528220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 312 and 642 of Human NDUFS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CI-75kD; CI-75Kd CI-75k PRO1304 NADH-ubiquinone oxidoreductase 75 kDa subunit mitochondrial complex I 75kDa subunit complex I mitochondrial respiratory chain 75-kD subunit mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit; complex I 75kDa subunit; complex I, mitochondrial respiratory chain, 75-kD subunit; Complex I-75kD; mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa; NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Q reductase); NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial
Gene Aliases: 5830412M15Rik; 9930026A05Rik; CI-75k; CI-75Kd; NDUFS1; PRO1304
UniProt ID: (Human) P28331, (Rat) Q66HF1, (Mouse) Q91VD9
Entrez Gene ID: (Human) 4719, (Rat) 301458, (Mouse) 227197
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.