Abnova
Manufacturer Code:PAB19219
Catalog # 89-120-749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids near C-terminus of human NDUFB9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of NDUFB9.
For Research Use Only
Protein Aliases: B22; CI-B22; complex I B22 subunit; Complex I-B22; DKFZp566O173; FLJ22885; LYR motif-containing protein 3; LYRM3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9; NADH-ubiquinone oxidoreductase B22 subunit; UQOR22
Gene Aliases: B22; CI-B22; LYRM3; NDUFB9; UQOR22
UniProt ID: (Human) Q9Y6M9
Entrez Gene ID: (Human) 4715
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.