Invitrogen
Manufacturer Code:PA529258
Catalog # PIPA529258
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 98 of Human NDUFB3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B12 CI-B12 NADH dehydrogenase &lsqbubiquinone] 1 beta subcomplex subunit 3 NADH-ubiquinone oxidoreductase B12 subunit complex I B12 subunit complex I-B12; CI-B12; Complex I-B12; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3; NADH-ubiquinone oxidoreductase B12 subunit
Gene Aliases: 2700033I16Rik; AI415450; Ndufb3
UniProt ID: (Mouse) Q9CQZ6
Entrez Gene ID: (Mouse) 66495
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.