Novus Biologicals
Manufacturer Code:NBP219504
Catalog # NBP219504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length human NDUFB3 Recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 (12kD B12) NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3 12kDa; CI-B12; Complex I-B12; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 3, 12kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3; NADH-ubiquinone oxidoreductase B12 subunit
Gene Aliases: B12; CI-B12; NDUFB3
UniProt ID: (Human) O43676
Entrez Gene ID: (Human) 4709
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.