Novus Biologicals
Manufacturer Code:NBP293366100UL
Catalog # NB028614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-98 of human NDUFB3 (NP_002482.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 (12kD B12) NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3 12kDa; CI-B12; Complex I-B12; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 3, 12kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3; NADH-ubiquinone oxidoreductase B12 subunit
Gene Aliases: B12; CI-B12; NDUFB3
UniProt ID: (Human) O43676
Entrez Gene ID: (Human) 4709
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.