Proteintech
Manufacturer Code:167201AP150UL
Catalog # 50-173-1136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NDUFB11 Fusion Protein Ag10159 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CI ESSS Complex I ESSS ESSS NDUFB11 Neuronal protein 17.3 P17.3; CI-ESSS; complex I NP17.3 subunit; Complex I-ESSS; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial; NADH-ubiquinone oxidoreductase ESSS subunit; Neuronal protein 15.6; Neuronal protein 17.3; Np15.6; Np17.3; nuclear protein 15.6; p15.6
Gene Aliases: CI-ESSS; D5Bwg0566e; D5Bwg0577e; ESSS; LSDMCA3; NDUFB11; Np15; NP15.6; NP17.3; P17.3; RGD1563698; UNQ111/PRO1064
UniProt ID: (Human) Q9NX14, (Mouse) O09111
Entrez Gene ID: (Human) 54539, (Rat) 299310, (Mouse) 104130
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.