Invitrogen
Manufacturer Code:PA5115630
Catalog # PIPA5115630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NDUFAF2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810058I14Rik B17.2L B17.2-like B172L B172-like C86051 mimitin mimitin mitochondrial MMTN Myc-induced mitochondria protein myc-induced mitochondrial protein NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 2 NADH dehydrogenase (ubiquinone) complex I assembly factor 2 NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2 NADH dehydrogenase 1 alpha subcomplex assembly factor 2 NADH:ubiquinone oxidoreductase complex assembly factor 2 NDUFA12L NDUFA12-like protein NDUFAF2 RGD1560158; B17.2-like; B17.2L; Mimitin; mimitin, mitochondrial; MMTN; Myc-induced mitochondria protein; Myc-induced mitochondrial protein; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2; NADH dehydrogenase (ubiquinone) complex I, assembly factor 2; NADH dehydrogenase 1 alpha subcomplex assembly factor 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2; NDUFA12-like protein; RGD1560158
Gene Aliases: 1810058I14Rik; B17.2L; C86051; mimitin; MMTN; NDUFA12L; NDUFAF2; RGD1560158
UniProt ID: (Human) Q8N183, (Mouse) Q59J78
Entrez Gene ID: (Human) 91942, (Rat) 361894, (Mouse) 75597
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.