Novus Biologicals
Manufacturer Code:NBP293263100UL
Catalog # NB028412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-169 of human NDUFAF2 (NP_777549.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B17.2-like; B17.2L; B17.2LFLJ22398 B17.2-like mimitin MMTNmitochondrial Myc-induced mitochondrial protein NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 2 NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2 NDUFA12-like NDUFA12-like protein; Mimitin; MMTN; Myc-induced mitochondrial protein; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2; NADH dehydrogenase (ubiquinone) complex I, assembly factor 2; NADH dehydrogenase 1 alpha subcomplex assembly factor 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2; NDUFA12-like protein
Gene Aliases: B17.2L; mimitin; MMTN; NDUFA12L; NDUFAF2
UniProt ID: (Human) Q8N183
Entrez Gene ID: (Human) 91942
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.