Invitrogen
Manufacturer Code:PA5103944
Catalog # PIPA5103944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NDUFA5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900002J19Rik B13 CI-13kB CI-13KD-B complex I 13kDa subunit B complex I subunit B13 Complex I-13kD-B NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5 NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5 NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5 13kDa NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5 NADH ubiquinone oxidoreductase subunit B13 NADH:ubiquinone oxidoreductase subunit A5 NADH-ubiquinone oxidoreductase 13 kDa-B subunit NADHUO NDUFA5 NUFM type I dehydrogenase ubiquinone reductase UQOR13; CI-13kD-B; complex I 13kDa subunit B; Complex I subunit B13; Complex I-13kD-B; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; type I dehydrogenase; ubiquinone reductase
Gene Aliases: 2900002J19Rik; B13; CI-13kB; CI-13KD-B; NDUFA5; NUFM; UQOR13
UniProt ID: (Human) Q16718, (Mouse) Q9CPP6
Entrez Gene ID: (Human) 4698, (Mouse) 68202
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.