Invitrogen
Manufacturer Code:PA572813
Catalog # PIPA572813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NDST2 antibody for a 16 amino acid peptide near the amino terminus of human NDST2. The immunogen is within amino acids 20 - 70 of NDST2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Heparan sulfate]-glucosamine N-sulfotransferase; Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 2; Glucosaminyl N-deacetylase/N-sulfotransferase 2; glucosaminyl N-deacetylase/N-sulphotransferase 2; heparan glucosaminyl N-deacetylase/N-sulfotransferase 2; heparan N-sulfotransferase 2; Heparan sulfate N-deacetylase 2; heparan sulfate N-deacetylase/N-sulfotransferase 2; Heparan sulfate N-sulfotransferase 2; HSST2 NST2 N-HSST 2 N-heparan sulfate sulfotransferase 2 NDST-2 bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 2 glucosaminyl N-deacetylase/N-sulfotransferase 2; mastocytoma N-deacetylase/N-sulfotransferase; Mndns; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2; N-deacetylase/N-sulfotransferase 2; N-heparan sulfate sulfotransferase 2; N-HSST 2; NDST-2
Gene Aliases: HSST2; Mndns; NDST-2; NDST2; NST2
UniProt ID: (Human) P52849, (Mouse) P52850
Entrez Gene ID: (Human) 8509, (Rat) 114002, (Mouse) 17423
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.