Invitrogen
Manufacturer Code:PA545611
Catalog # PIPA545611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NDRG4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain development-related molecule 1; development-related protein; N-myc downstream regulated 4; N-myc downstream regulated gene 4; N-myc downstream-regulated gene 4 protein; NDRG family member 4 DKFZp686I1615 FLJ30586 FLJ42011 KIAA1180 MGC19632 SMAP-8; Protein NDRG4; SMAP-8; smooth muscle-associated protein 8; Vascular smooth muscle cell-associated protein 8
Gene Aliases: BDM1; KIAA1180; Ndr4; NDRG4; SMAP-8; SMAP8
UniProt ID: (Human) Q9ULP0
Entrez Gene ID: (Pig) 100513927, (Human) 65009, (Rat) 64457
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.