Novus Biologicals
Manufacturer Code:NBP155431
Catalog # NBP155431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NDRG2(NDRG family member 2) The peptide sequence was selected from the C terminal of NDRG2. Peptide sequence GYMASSCMTRLSRSRTASLTSAASVDGNRSRSRTLSQSSESGTLSSGPPG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytoplasmic protein Ndr1; DKFZp781G1938 FLJ25522 KIAA1248cytoplasmic protein Ndr1 NDR1-related protein NDR2 NDRG family member 2 protein NDRG2 Protein Syld709613 syld709613 protein SYLDN-myc downstream regulator 2; N-myc downstream regulator 2; N-myc downstream-regulated gene 2 protein; NDR1-related protein NDR2; Protein NDRG2; Protein Syld709613; syld709613 protein
Gene Aliases: KIAA1248; NDRG2; SYLD
UniProt ID: (Human) Q9UN36
Entrez Gene ID: (Human) 57447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.