Invitrogen
Manufacturer Code:PA523230
Catalog # PIPA523230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (382 GNSAGPKSMEVSC 394) of NDRG1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP43 CMT4D DRG1 GC4 HMSNL NDR1 NMSL PROXY1 RIT42 RTP TARG1 TDD5 DRG-1 N-myc downstream-regulated gene 1 protein differentiation-related gene 1 protein nickel-specific induction protein Cap43 protein NDRG1 protein regulated by oxygen-1 reducing agents and tunicamycin-responsive protein tunicamycin-responsive protein; Differentiation-related gene 1 protein; DRG-1; N-myc downstream regulated 1; N-myc downstream-regulated gene 1 protein; Nickel-specific induction protein Cap43; Protein Ndr1; Protein NDRG1; protein regulated by oxygen-1; Reducing agents and tunicamycin-responsive protein; Rit42; RTP
Gene Aliases: CAP43; CMT4D; DRG-1; DRG1; GC4; HMSNL; NDR1; NDRG1; Ndrl; NMSL; PROXY1; RIT42; RTP; TARG1; TDD5
UniProt ID: (Human) Q92597, (Mouse) Q62433
Entrez Gene ID: (Human) 10397, (Mouse) 17988
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.