Novus Biologicals
Manufacturer Code:NBP223658SS
Catalog # NBP223658SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acid 350-400 of human NDRG1 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP43 CMT4D Differentiation-related gene 1 protein DRG-1 DRG1HMSNL NDR1 Nickel-specific induction protein Cap43 NMSL N-myc downstream regulated 1 N-myc downstream-regulated gene 1 protein protein NDRG1 protein regulated by oxygen-1 PROXY1 Reducing agents and tunicamycin-responsive protein RIT42 RTPGC4 TARG1 TDD5 tunicamycin-responsive protein; Differentiation-related gene 1 protein; DRG-1; N-myc downstream-regulated gene 1 protein; Nickel-specific induction protein Cap43; Protein NDRG1; protein regulated by oxygen-1; Reducing agents and tunicamycin-responsive protein; Rit42; RTP
Gene Aliases: CAP43; CMT4D; DRG-1; DRG1; GC4; HMSNL; NDR1; NDRG1; NMSL; PROXY1; RIT42; RTP; TARG1; TDD5
UniProt ID: (Human) Q92597
Entrez Gene ID: (Human) 10397
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.