Proteintech
Manufacturer Code:269021AP150UL
Catalog # 50-173-5495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NDRG1 Fusion Protein Ag25424 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP43 CMT4D DRG 1 DRG1 GC4 HMSNL N myc downstream regulated 1 NDR1 NDRG1 Protein NDRG1 PROXY1 RIT42 RTP TARG1 TDD5; Differentiation-related gene 1 protein; DRG-1; N-myc downstream-regulated gene 1 protein; Nickel-specific induction protein Cap43; Protein NDRG1; protein regulated by oxygen-1; Reducing agents and tunicamycin-responsive protein; Rit42; RTP
Gene Aliases: CAP43; CMT4D; DRG-1; DRG1; GC4; HMSNL; NDR1; NDRG1; NMSL; PROXY1; RIT42; RTP; TARG1; TDD5
UniProt ID: (Human) Q92597
Entrez Gene ID: (Human) 10397
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.