Invitrogen
Manufacturer Code:PA5102534
Catalog # PIPA5102534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NDP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EVR2 FEVR ND Ndp NDP norrin cystine knot growth factor Ndph Norrie disease (pseudoglioma) Norrie disease (pseudoglioma) (human) Norrie disease homolog norrie disease protein Norrie disease protein homolog Norrin RGD1563968 X-linked exudative vitreoretinopathy 2 protein; Norrie disease (pseudoglioma); Norrie disease homolog; Norrie disease protein; Norrie disease protein homolog; Norrin; X-linked exudative vitreoretinopathy 2 protein
Gene Aliases: EVR2; FEVR; ND; NDP; Ndph; RGD1563968
UniProt ID: (Human) Q00604, (Mouse) P48744
Entrez Gene ID: (Human) 4693, (Mouse) 17986, (Rat) 363443
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.