Novus Biologicals
Manufacturer Code:NBP272896R
Catalog # NBP272896R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E3 |
Immunogen | Full length recombinant protein of human NDOR1 (NP_055249) produced in HEK293T cells. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: bA350O14.9 EC 1.6 MGC138148 NADPH dependent diflavin oxidoreductase 1 NADPH-dependent diflavin oxidoreductase 1 NADPH-dependent FMN and FAD containing oxidoreductase NADPH-dependent FMN and FAD-containing oxidoreductase Novel reductase 1 NR1NADPH dependent FMN and FAD containing oxidoreductase; NADPH-dependent diflavin oxidoreductase 1; NADPH-dependent FMN and FAD-containing oxidoreductase; NDOR1; Novel reductase 1
Gene Aliases: bA350O14.9; NDOR1; NR1
UniProt ID: (Human) Q9UHB4
Entrez Gene ID: (Human) 27158
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.