OriGene
Manufacturer Code:CF505464
Catalog # 50-166-3657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E3 |
Immunogen | Full length human recombit protein of human NDOR1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NADPH-dependent diflavin oxidoreductase 1; NADPH-dependent FMN and FAD-containing oxidoreductase; NDOR1; Novel reductase 1; NR1 bA350O14.9
Gene Aliases: 4930447P04Rik; bA350O14.9; Ndor; NDOR1; NR1
UniProt ID: (Human) Q9UHB4, (Mouse) A2AI05
Entrez Gene ID: (Human) 27158, (Dog) 491234, (Mouse) 78797
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.