Invitrogen
Manufacturer Code:MA518029
Catalog # PIMA518029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | N1Nterm1532.8 |
Immunogen | N-terminal peptide of murine Ndfip 1 coupled to KLH (VEPACGSGYQQLQNEEPGE) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: NDFIP1 NEDD4 family-interacting protein 1 N4WBP5 NEDD4 WW domain-binding protein 5 N4wbp5; NEDD4 family-interacting protein 1; Nedd4 WW binding protein 5; NEDD4 WW domain-binding protein 5; Nedd4 WW-binding protein 5
Gene Aliases: 0610010M22Rik; N4wbp5; Ndfip1
UniProt ID: (Mouse) Q8R0W6
Entrez Gene ID: (Mouse) 65113
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.