Invitrogen
Manufacturer Code:PA5113477
Catalog # PIPA5113477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human NEDD4 family-interacting protein 1 protein (2-116AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610010M22Rik Breast cancer-associated protein SGA-1M N4WBP5 NDFIP1 Nedd4 family interacting protein 1 NEDD4 family-interacting protein 1 Nedd4 WW binding protein 5 NEDD4 WW domain-binding protein 5 Nedd4 WW-binding protein 5 PSEC0192 PSEC0223 Putative MAPK-activating protein PM13 Putative NF-kappa-B-activating protein 164 Putative NFKB and MAPK-activating protein; Breast cancer-associated protein SGA-1M; NEDD4 family-interacting protein 1; NEDD4 WW domain-binding protein 5; Putative MAPK-activating protein PM13; Putative NF-kappa-B-activating protein 164; Putative NFKB and MAPK-activating protein
Gene Aliases: N4WBP5; NDFIP1; PSEC0192; PSEC0223
UniProt ID: (Human) Q9BT67
Entrez Gene ID: (Human) 80762
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.