Invitrogen
Manufacturer Code:PA5101992
Catalog # PIPA5101992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NDFIP1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610010M22Rik Breast cancer-associated protein SGA-1M N4WBP5 Ndfip1 Nedd4 family interacting protein 1 NEDD4 family-interacting protein 1 Nedd4 WW binding protein 5 NEDD4 WW domain-binding protein 5 Nedd4 WW-binding protein 5 PSEC0192 PSEC0223 Putative MAPK-activating protein PM13 putative NF-kappa-B-activating protein 164 Putative NFKB and MAPK-activating protein; Breast cancer-associated protein SGA-1M; NEDD4 family-interacting protein 1; Nedd4 WW binding protein 5; NEDD4 WW domain-binding protein 5; Nedd4 WW-binding protein 5; Putative MAPK-activating protein PM13; Putative NF-kappa-B-activating protein 164; Putative NFKB and MAPK-activating protein
Gene Aliases: 0610010M22Rik; N4WBP5; NDFIP1; PSEC0192; PSEC0223
UniProt ID: (Human) Q9BT67, (Rat) Q5U2S1, (Mouse) Q8R0W6
Entrez Gene ID: (Human) 80762, (Rat) 291609, (Mouse) 65113
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.