Proteintech
Manufacturer Code:172621AP150UL
Catalog # 50-173-1475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NDEL1 Fusion Protein Ag10975 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endooligopeptidase A; EOPA MITAP1 Mitosin associated protein 1 NDEL1 NUDEL Protein Nudel; LIS1-interacting protein NUDEL; LIS1-interacting protein NUDEL; endooligopeptidase A; Mitosin-associated protein 1; mitosin-associated protein MITAP1; mNudE-L; nuclear disribution gene E like; nuclear distribution gene E-like homolog 1; Nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog like 1; nudE nuclear distribution gene E homolog-like 1; Protein mNudE-like; Protein Nudel
Gene Aliases: 2600006O07Rik; EOPA; MITAP1; mNudel; NDE1L1; NDE2; NDEL1; NUDE2; NUDEL
UniProt ID: (Human) Q9GZM8, (Rat) Q78PB6, (Mouse) Q9ERR1
Entrez Gene ID: (Human) 81565, (Rat) 170845, (Mouse) 83431
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.