Novus Biologicals
Manufacturer Code:NBP222627
Catalog # NBP222627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F7 |
Immunogen | Recombinant human NDE1 (1-335aa) purified from E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: FLJ20101 HOM-TES-87 nuclear distribution protein nudE homolog 1 NudE nudE nuclear distribution gene E homolog 1 (A. nidulans) NUDE1 rat homolog; LIS1-interacting protein NUDE1, rat homolog; Nuclear distribution protein nudE homolog 1; NudE; nudE nuclear distribution E homolog 1; nudE nuclear distribution gene E homolog 1
Gene Aliases: HOM-TES-87; LIS4; MHAC; NDE; NDE1; NUDE; NUDE1
UniProt ID: (Human) Q9NXR1
Entrez Gene ID: (Human) 54820
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.