Invitrogen
Manufacturer Code:PA531121
Catalog # PIPA531121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 203 of Human NDE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HOM-TES-87 LIS4 NUDE NUDE1 LIS1-interacting protein NUDE1 rat homolog nuclear distribution protein nudE homolog 1; LIS1-interacting protein NUDE1, rat homolog; Nuclear distribution protein nudE homolog 1; NudE; nudE nuclear distribution E homolog 1; nudE nuclear distribution gene E homolog 1
Gene Aliases: HOM-TES-87; LIS4; MHAC; NDE; NDE1; NUDE; NUDE1
UniProt ID: (Human) Q9NXR1
Entrez Gene ID: (Human) 54820
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.