Novus Biologicals
Manufacturer Code:NBP297461UV
Catalog # NB043828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human NCOR1 fragment. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hN-CoR KIAA1047N-CoR1 MGC104216 N-CoRhCIT529I10 nuclear receptor corepressor 1 nuclear receptor co-repressor 1 thyroid hormone- and retinoic acid receptor-associated corepressor 1 TRAC1; N-CoR; Nuclear receptor corepressor 1; protein phosphatase 1, regulatory subunit 109; thyroid hormone- and retinoic acid receptor-associated corepressor 1
Gene Aliases: hN-CoR; KIAA1047; N-CoR; N-CoR1; NCOR1; PPP1R109; TRAC1
UniProt ID: (Human) O75376
Entrez Gene ID: (Human) 9611
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.