Proteintech
Manufacturer Code:105561AP150UL
Catalog # 50-172-6403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NCL Fusion Protein Ag0859 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C23 NCL nucleolin Protein C23; Nucleolin; nucleolin-like protein; Protein C23
Gene Aliases: B530004O11Rik; C23; D0Nds28; D1Nds28; NCL; Nuc; Nucl
UniProt ID: (Human) P19338, (Mouse) P09405
Entrez Gene ID: (Human) 4691, (Mouse) 17975, (Rat) 25135
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.