Invitrogen
Manufacturer Code:PA558813
Catalog # PIPA558813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NCKAP1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hematopoietic protein 1; Membrane-associated protein HEM-1; Nck-associated protein 1-like; Nck-associated protein 1-like Hematopoietic protein 1 Membrane-associated protein HEM-1 NCKAP1L HEM1
Gene Aliases: HEM1; NCKAP1L
UniProt ID: (Human) P55160
Entrez Gene ID: (Human) 3071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.