Invitrogen
Manufacturer Code:PA521680
Catalog # PIPA521680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 693 and 936 of NCKAP1 (Uniprot IDNo.Q9Y2A7) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain protein H19; HEM2 NAP1 NAP125 NAP 1 membrane-associated protein HEM-2 nck-associated protein 1 p125Nap1; Membrane-associated protein HEM-2; MH19; NAP 1; Nck-associated protein 1; p125Nap1
Gene Aliases: C79304; H19; Hem-2; HEM2; KIAA0587; mh19; mKIAA0587; NAP1; NAP125; NCKAP1; p125Nap1
UniProt ID: (Human) Q9Y2A7, (Mouse) P28660, (Rat) P55161
Entrez Gene ID: (Human) 10787, (Mouse) 50884, (Rat) 58823
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.