Invitrogen
Manufacturer Code:PA529210
Catalog # PIPA529210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 286 of Human Nck beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic protein NCK2; GRB4 NCKbeta SH2/SH3 adaptor protein NCK-beta cytoplasmic protein NCK2 growth factor receptor-bound protein 4 noncatalytic region of tyrosine kinase beta; Growth factor receptor-bound protein 4; NCK adaptor protein 2; Nck-2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta
Gene Aliases: GRB4; NCK2; NCKbeta
UniProt ID: (Human) O43639
Entrez Gene ID: (Human) 8440
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.