Invitrogen
Manufacturer Code:PA5112625
Catalog # PIPA5112625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Cytoplasmic protein NCK2 protein (1-380AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4833426I10Rik Cytoplasmic protein NCK2 GRB4 Growth factor receptor-bound protein 4 NCK adaptor protein 2 NCK2 Nck-2 NCKbeta non-catalytic region of tyrosine kinase adaptor protein 2 noncatalytic region of tyrosine kinase beta SH2/SH3 adaptor protein NCK-beta; Cytoplasmic protein NCK2; Growth factor receptor-bound protein 4; NCK adaptor protein 2; Nck-2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta
Gene Aliases: 4833426I10Rik; GRB4; Nck-2; NCK2; NCKbeta
UniProt ID: (Human) O43639, (Mouse) O55033
Entrez Gene ID: (Human) 8440, (Mouse) 17974
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.