Novus Biologicals
Manufacturer Code:NBP29350320UL
Catalog # NB028883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-380 of human NCK2 (NP_003572.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic protein NCK2; GRB4 NCK adaptor protein 2cytoplasmic protein NCK2 Nck-2 NCKbeta noncatalytic region of tyrosine kinase beta SH2/SH3 adaptor protein NCK-beta; Growth factor receptor-bound protein 4; NCK adaptor protein 2; Nck-2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta
Gene Aliases: GRB4; NCK2; NCKbeta
UniProt ID: (Human) O43639
Entrez Gene ID: (Human) 8440
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.