Invitrogen
Manufacturer Code:PA599120
Catalog # PIPA599120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit human protein purified from E.coli (HIS-NCK2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic protein NCK1; Cytoplasmic protein NCK2; Growth factor receptor-bound protein 4; melanoma NCK protein; NCK adaptor protein 1; NCK adaptor protein 2; NCK tyrosine kinase; Nck-1; Nck-2; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-alpha; SH2/SH3 adaptor protein NCK-beta
Gene Aliases: 4833426I10Rik; 6330586M15Rik; D230010O13Rik; GRB4; NCK; Nck-1; Nck-2; NCK1; NCK2; NCKalpha; NCKbeta
UniProt ID: (Human) P16333, (Human) O43639, (Mouse) Q99M51, (Mouse) O55033
Entrez Gene ID: (Human) 4690, (Human) 8440, (Rat) 300955, (Mouse) 17973, (Mouse) 17974, (Rat) 316369
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.